Skip to Content
MilliporeSigma
All Photos(1)

Documents

617466

Sigma-Aldrich

Acetyl-d3-L-carnitine hydrochloride

≥98 atom % D, ≥99% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9D3H14NO4·HCl
CAS Number:
Molecular Weight:
242.72
UNSPSC Code:
12352108
PubChem Substance ID:

isotopic purity

≥98 atom % D

Assay

≥99% (CP)

form

solid

optical activity

[α]25/D -28°, c = 1 in H2O

mp

194 °C (dec.) (lit.)

mass shift

M+3

storage temp.

−20°C

SMILES string

[Cl-].[2H]C([2H])([2H])C(=O)O[C@H](CC(O)=O)C[N+](C)(C)C

InChI

1S/C9H17NO4.ClH/c1-7(11)14-8(5-9(12)13)6-10(2,3)4;/h8H,5-6H2,1-4H3;1H/t8-;/m1./s1/i1D3;

InChI key

JATPLOXBFFRHDN-SPMMGKNWSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna Marco-Ramell et al.
Journal of proteome research, 17(7), 2307-2317 (2018-06-16)
This study explores the metabolic profiles of concordant/discordant phenotypes of high insulin resistance (IR) and obesity. Through untargeted metabolomics (LC-ESI-QTOF-MS), we analyzed the fasting serum of subjects with high IR and/or obesity ( n = 64). An partial least-squares discriminant
Ximena Mora-Cubillos et al.
Molecular nutrition & food research, 59(12), 2480-2490 (2015-09-29)
To identify the most discriminant dietary biomarkers of nuts exposure in subjects with metabolic syndrome (MetS), and investigate the potential association between exposure and the severity of the MetS diagnostic traits. We applied the untargeted LC-ESI-qToF-MS-driven metabolomic workflow to explore

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service