Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

617075

Sigma-Aldrich

Diethylamine-d11

98 atom % D, 98% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CD3CD2)2ND
CAS Number:
Molecular Weight:
84.20
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

isotopic purity

98 atom % D

Assay

98% (CP)

bp

55 °C (lit.)

mp

-50 °C (lit.)

density

0.811 g/mL at 25 °C

mass shift

M+11

SMILES string

[2H]N(C([2H])([2H])C([2H])([2H])[2H])C([2H])([2H])C([2H])([2H])[2H]

InChI

1S/C4H11N/c1-3-5-4-2/h5H,3-4H2,1-2H3/i1D3,2D3,3D2,4D2/hD

InChI key

HPNMFZURTQLUMO-REUZILGJSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service