Skip to Content
MilliporeSigma
All Photos(1)

Documents

615749

Sigma-Aldrich

Sodium propionate-d5

98 atom % D

Synonym(s):

Propionic-d5 acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3CD2CO2Na
CAS Number:
Molecular Weight:
101.09
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

98 atom % D

form

solid

mp

285-286 °C (lit.)

mass shift

M+5

SMILES string

[Na+].[2H]C([2H])([2H])C([2H])([2H])C([O-])=O

InChI

1S/C3H6O2.Na/c1-2-3(4)5;/h2H2,1H3,(H,4,5);/q;+1/p-1/i1D3,2D2;

InChI key

JXKPEJDQGNYQSM-LUIAAVAXSA-M

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas W Hoffmann et al.
The ISME journal, 10(2), 460-477 (2015-07-29)
Studying host-microbiota interactions are fundamental to understanding the mechanisms involved in intestinal homeostasis and inflammation. In this work, we analyzed these interactions in mice that were mono-associated with six microorganisms that are representative of inflammatory bowel disease (IBD)-associated dysbiosis: the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service