Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

615706

Sigma-Aldrich

Butyric acid-4,4,4-d3

98 atom % D

Synonym(s):

Butanoic-4,4,4-d3 acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3CH2CH2CO2H
CAS Number:
Molecular Weight:
91.12
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

98 atom % D

Quality Level

Assay

99% (CP)

bp

162 °C (lit.)

mp

-6--3 °C (lit.)

density

0.996 g/mL at 25 °C

mass shift

M+3

SMILES string

[2H]C([2H])([2H])CCC(O)=O

InChI

1S/C4H8O2/c1-2-3-4(5)6/h2-3H2,1H3,(H,5,6)/i1D3

InChI key

FERIUCNNQQJTOY-FIBGUPNXSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

161.6 °F - closed cup

Flash Point(C)

72 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Meike Kespohl et al.
Frontiers in immunology, 8, 1036-1036 (2017-09-13)
Short-chain fatty acids (SCFAs), which are generated by the bacterial fermentation of dietary fibers, promote expansion of regulatory T cells (Tregs). Potential therapeutic value of SCFAs has been recently highlighted in the experimental models of T cell-mediated autoimmunity and allergic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service