Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

615218

Sigma-Aldrich

Triphenyl phosphate-d15

98 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6D5O)3PO
Molecular Weight:
341.38
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:

isotopic purity

98 atom % D

form

solid

bp

244 °C/10 mmHg (lit.)

mp

48-50 °C (lit.)

mass shift

M+15

SMILES string

[2H]c1c([2H])c([2H])c(OP(=O)(Oc2c([2H])c([2H])c([2H])c([2H])c2[2H])Oc3c([2H])c([2H])c([2H])c([2H])c3[2H])c([2H])c1[2H]

InChI

1S/C18H15O4P/c19-23(20-16-10-4-1-5-11-16,21-17-12-6-2-7-13-17)22-18-14-8-3-9-15-18/h1-15H/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D,11D,12D,13D,14D,15D

InChI key

XZZNDPSIHUTMOC-KLHTYYPYSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yeowool Choi et al.
The Science of the total environment, 708, 135106-135106 (2019-12-04)
Triphenyl phosphate (TPHP), one of the organophosphate flame retardants, has been widely used in manufacturing, thereby causing a gradual increase in TPHP concentrations in aquatic environments. However, the information on the biotransformation mechanism of TPHP in invertebrates is lacking. The
Florence Nantaba et al.
Chemosphere, 262, 127716-127716 (2020-08-18)
For the first time the occurrence of 25 organic micropollutants (OMPs) including; 11 personal care products (PCPs), six phthalate ester plasticizers (PEPs) and eight organophosphorus flame retardants (OPFRs) was investigated in 72 water samples obtained from five bays in the
Long Pang et al.
Environmental science and pollution research international, 26(27), 27862-27871 (2019-07-26)
In this study, nine OPEs were measured in 60 street dust samples collected from the central province of Henan, China. The total concentrations of OPEs (∑9OPEs) in street dust range from 2.77 to 505 μg/kg (median 59.3 μg/kg). Chlorinated OPEs are dominant
Nele Van den Eede et al.
Toxicology in vitro : an international journal published in association with BIBRA, 29(8), 2045-2054 (2015-09-01)
Since the publication of REACH guidelines, the need for in vitro tools for toxicity testing has increased. We present here the development of a hepatotoxicity testing tool using human HepaRG cell cultures and metabolomics. HepaRG cells were exposed to either

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service