Sign In to View Organizational & Contract Pricing.
About This Item
Empirical Formula (Hill Notation):
C7D6H3N
CAS Number:
Molecular Weight:
113.19
UNSPSC Code:
12352005
PubChem Substance ID:
MDL number:
Isotopic purity:
98 atom % D
Mass shift:
M+6
isotopic purity
98 atom % D
bp
143-145 °C
density
0.971 g/mL at 25 °C
mass shift
M+6
SMILES string
[2H]C([2H])([2H])c1cccc(n1)C([2H])([2H])[2H]
InChI
1S/C7H9N/c1-6-4-3-5-7(2)8-6/h3-5H,1-2H3/i1D3,2D3
InChI key
OISVCGZHLKNMSJ-WFGJKAKNSA-N
General description
2,6-Lutidine-(dimethyl-d6) 98 atom % D is a high-purity isotope product with a unique chemical profile that belongs to the class of isotopes. Produced by Sigma-Aldrich, we are committed to providing quality isotope products and a reliable supply chain. Available in industrial and pre-pack quantities.
Application
- Pharmaceutical formulation development
- Mass spectrometry calibration
- reaction mechanism investigations
- Analytical method development
- Mass spectrometry calibration
- reaction mechanism investigations
- Analytical method development
Features and Benefits
Features
Benefits
- High purity of 2,6-Lutidine-(dimethyl-d6) 98 atom % D ensures optimal performance.
- Cost-effective solution for 2,6-Lutidine-(dimethyl-d6) 98 atom % D applications.
- Exceptional stability and compatibility with various ingredients.
- Versatile use in different formulations involving 2,6-Lutidine-(dimethyl-d6) 98 atom % D.
Benefits
- Increases market competitiveness of products containing 2,6-Lutidine-(dimethyl-d6) 98 atom % D.
- Enhances product efficacy and reliability in formulations.
- Reduces time to market for products utilizing 2,6-Lutidine-(dimethyl-d6) 98 atom % D.
- Improves overall product quality and consumer satisfaction.
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
wgk
WGK 3
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
3 - Flammable liquids
flash_point_f
98.6 °F - closed cup
flash_point_c
37 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

