Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

609838

Sigma-Aldrich

2,2-Dimethylpropanol-17O

20 atom % 17O

Synonym(s):

Neopentyl alcohol-17O

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CCH217OH
Molecular Weight:
89.15
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

isotopic purity

20 atom % 17O

mol wt

88.36 by atom % calculation (20 atom % 17O)

bp

113-114 °C (lit.)

mp

52-56 °C (lit.)

density

0.820 g/mL at 25 °C

mass shift

M+1

SMILES string

CC(C)(C)C[17OH]

InChI

1S/C5H12O/c1-5(2,3)4-6/h6H,4H2,1-3H3/i6+1

InChI key

KPSSIOMAKSHJJG-PTQBSOBMSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service