Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

607517

Sigma-Aldrich

Acetyl chloride-1-13C,d3

98 atom % D, 99 atom % 13C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD313COCl
CAS Number:
Molecular Weight:
82.51
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

99 atom % 13C
98 atom % D

Assay

99% (CP)

technique(s)

protein expression: suitable

bp

52 °C (lit.)

mp

-112 °C (lit.)

density

1.160 g/mL at 25 °C

mass shift

M+4

storage temp.

2-8°C

SMILES string

Cl[13C](C([2H])([2H])[2H])=O

InChI

1S/C2H3ClO/c1-2(3)4/h1H3/i1D3,2+1

InChI key

WETWJCDKMRHUPV-HZPPXAECSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

41.0 °F

Flash Point(C)

5 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service