Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

605832

Sigma-Aldrich

Octanoic acid-1-13C Extra

99 atom % 13C

Synonym(s):

Octanoic acid-1-13C, Caprylic acid-1-13C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)613CO2H
CAS Number:
Molecular Weight:
145.20
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

description

Special high CP

isotopic purity

99 atom % 13C

refractive index

n20/D 1.4284 (lit.)

bp

237 °C (lit.)

mp

16 °C (lit.)

density

0.916 g/mL at 25 °C

mass shift

M+1

SMILES string

CCCCCCC[13C](O)=O

InChI

1S/C8H16O2/c1-2-3-4-5-6-7-8(9)10/h2-7H2,1H3,(H,9,10)/i8+1

InChI key

WWZKQHOCKIZLMA-VJJZLTLGSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1C

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service