Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

605506

Sigma-Aldrich

D-Glucose-2,5-13C2

99 atom % 13C

Synonym(s):

Labeled Glucose, Dextrose-2,5-13C2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C2C4H12O6
Molecular Weight:
182.14
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

isotopic purity

99 atom % 13C

Assay

99% (CP)

form

powder

optical activity

[α]25/D +52.0°, c = 2 in H2O (trace NH4OH)

mp

150-152 °C (lit.)

mass shift

M+2

SMILES string

OC[13C@H]1OC(O)[13C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5-,6?/m1/s1/i2+1,5+1

InChI key

WQZGKKKJIJFFOK-BUFNXKTDSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Isotopic Labeling for NMR Spectroscopy of Biological Solids;

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service