Skip to Content
MilliporeSigma
All Photos(1)

Documents

59539

Sigma-Aldrich

(4S,5S)-2,2-Dimethyl-1,3-dioxolane-4,5-dimethanol

≥97.0% (sum of enantiomers, GC)

Synonym(s):

(+)-2,3-O-Isopropylidene-L-threitol, (4S,5S)-4,5-Bis(hydroxymethyl)-2,2-dimethyl-1,3-dioxolane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H14O4
CAS Number:
Molecular Weight:
162.18
Beilstein:
1280797
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97.0% (sum of enantiomers, GC)

form

solid

optical activity

[α]20/D +3.0±0.5°, c = 5% in ethanol

bp

92-94 °C/0.1 mmHg (lit.)

mp

46-50 °C

SMILES string

CC1(C)O[C@@H](CO)[C@H](CO)O1

InChI

1S/C7H14O4/c1-7(2)10-5(3-8)6(4-9)11-7/h5-6,8-9H,3-4H2,1-2H3/t5-,6-/m0/s1

InChI key

INVRLGIKFANLFP-WDSKDSINSA-N

Application

(4S,5S)-2,2-Dimethyl-1,3-dioxolane-4,5-dimethanol can be used:
  • As a key intermediate for the preparation of (6Z)-cis-9S,10R-epoxy-nonadecene, a female sex pheromone of elm spanworm.
  • To prepare a chiral diphosphite ligand by reacting with phosphorochloridite using excess of NEt3. 

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of chiral alkenyl epoxides: the sex pheromone of the elm spanworm Ennomus subsignaria (Hubner)(Lepidoptera: Geometridae)
MaGee DI, et al.
Tetrahedron, 67(29), 5329-5338 (2011)
Modular chiral diphosphite derived from l-tartaric acid. Applications in metal-catalyzed asymmetric reactions
Rosas-Hern'andez A, et al.
J. Mol. Catal. A: Chem., 328(1-2), 68-75 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service