Skip to Content
MilliporeSigma
All Photos(3)

Documents

594636

Sigma-Aldrich

Hafnium(IV) carbide

<1.25 μm particle size

Synonym(s):

Hafnium carbide, Hafnium monocarbide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HfC
CAS Number:
Molecular Weight:
190.50
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

powder

Quality Level

reaction suitability

core: hafnium
reagent type: catalyst

particle size

<1.25 μm

mp

2930 °C (lit.)

density

12.2 g/mL at 25 °C (lit.)

SMILES string

[C-]#[Hf+]

InChI

1S/C.Hf/q-1;+1

InChI key

NVDNLVYQHRUYJA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Used for preparation of ultra-high temperature ceramics
  • Reactant in synthesis of hafnium-containing organometallic polymers
  • Additive in alloys
  • Used in coatings

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Sol. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service