Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

589861

Sigma-Aldrich

4-Methoxybenzaldehyde-α-d1

98 atom % D, 96% (CP)

Synonym(s):

p-Anisaldehyde-α-d1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8DH7O2
CAS Number:
Molecular Weight:
137.15
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

98 atom % D

Quality Level

Assay

96% (CP)

bp

248 °C (lit.)

density

1.127 g/mL at 25 °C

mass shift

M+1

SMILES string

[2H]C(=O)c1ccc(OC)cc1

InChI

1S/C8H8O2/c1-10-8-4-2-7(6-9)3-5-8/h2-6H,1H3/i6D

InChI key

ZRSNZINYAWTAHE-RAMDWTOOSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service