Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

586803

Sigma-Aldrich

1-(3-Aminophenyl)acetylene-1-13C

99 atom % 13C

Synonym(s):

3-(ethynyl-1)Benzenamine-13C

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13CC7H7N
CAS Number:
Molecular Weight:
118.14
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

isotopic purity

99 atom % 13C

bp

78-80 °C/2 mmHg (lit.)

density

1.049 g/mL at 25 °C

mass shift

M+1

SMILES string

Nc1cccc(c1)[13C]#C

InChI

1S/C8H7N/c1-2-7-4-3-5-8(9)6-7/h1,3-6H,9H2/i2+1

InChI key

NNKQLUVBPJEUOR-VQEHIDDOSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

138.2 °F - closed cup

Flash Point(C)

59 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service