Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

577820

Sigma-Aldrich

Acrylamide-1-13C

99 atom % 13C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CH13CONH2
CAS Number:
Molecular Weight:
72.07
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

isotopic purity

99 atom % 13C

form

solid

technique(s)

protein expression: suitable

bp

125 °C/25 mmHg (lit.)

mp

82-86 °C (lit.)

mass shift

M+1

SMILES string

N[13C](=O)C=C

InChI

1S/C3H5NO/c1-2-3(4)5/h2H,1H2,(H2,4,5)/i3+1

InChI key

HRPVXLWXLXDGHG-LBPDFUHNSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Carc. 1B - Eye Irrit. 2 - Muta. 1B - Repr. 2 - Resp. Sens. 1 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service