Skip to Content
MilliporeSigma
All Photos(1)

Documents

570257

Sigma-Aldrich

Fluorescein O,O′-diacrylate

98%

Synonym(s):

3’,6′-Diacryloxyspirobenzo[c]-furan[1,9’]xanthen-3-one, Diacryloyloxyfluorescein

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H16O7
CAS Number:
Molecular Weight:
440.40
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

solid

mp

160-163 °C (lit.)

λmax

203 nm

storage temp.

2-8°C

SMILES string

C=CC(=O)Oc1ccc2c(Oc3cc(OC(=O)C=C)ccc3C24OC(=O)c5ccccc45)c1

InChI

1S/C26H16O7/c1-3-23(27)30-15-9-11-19-21(13-15)32-22-14-16(31-24(28)4-2)10-12-20(22)26(19)18-8-6-5-7-17(18)25(29)33-26/h3-14H,1-2H2

InChI key

GTQZGVDHWVBMOJ-UHFFFAOYSA-N

Application

Fluorescent monomer

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alejandra Gonzalez-Urias et al.
Colloids and surfaces. B, Biointerfaces, 182, 110365-110365 (2019-07-26)
Stimuli-responsive polymeric nanogels have been proposed as nanocarriers of cisplatin to maximize its effect for cancer treatment. In this work, a comparative study between anionic core nanogels (ACN) and cationic core nanogels (CCN), both with PEGylated shells, has been performed.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service