Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

567663

Sigma-Aldrich

Ethyl cis-(β-cyano)acrylate

97%

Synonym(s):

Ethyl (Z )-3-cyano-2-propenoate, Ethyl cis -β-cyanoacrylate, Ethyl cis-β-cyanoacrylate, Ethyl cis-3-cyanoacrylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NCCH=CHCO2C2H5
CAS Number:
Molecular Weight:
125.13
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

refractive index

n20/D 1.4530 (lit.)

bp

215 °C (lit.)

density

1.044 g/mL at 25 °C (lit.)

SMILES string

[H]\C(C#N)=C(/[H])C(=O)OCC

InChI

1S/C6H7NO2/c1-2-9-6(8)4-3-5-7/h3-4H,2H2,1H3/b4-3-

InChI key

AMQZZSZCLSVKLO-ARJAWSKDSA-N

Application

Ethyl cis-β-cyanoacrylate behaves as a dienophile in the synthesis of indeno[2,1-a]pyrrolo[3,4-c]carbazole lactam regioisomers via Diels-Alder reaction.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

225.0 °F - closed cup

Flash Point(C)

107.2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of indeno [2, 1?a] pyrrolo [3, 4?c] carbazole lactam regioisomers using ethyl cis???cyanoacrylate as a dienophile and lactam precursor.
Hudkins RL and Park CH
Journal of Heterocyclic Chemistry, 40(1), 135-142 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service