559873
Dess-Martin periodinane solution
0.3 M in methylene chloride
Synonym(s):
1,1,1-Triacetoxy-1,1-dihydro-1,2-benziodoxol-3(1H)-one, Dess-Martin reagent, Triacetoxyperiodinane
About This Item
Recommended Products
Quality Level
reaction suitability
reagent type: oxidant
concentration
0.3 M in methylene chloride
bp
40 °C
density
1.369 g/mL at 25 °C
functional group
iodo
storage temp.
2-8°C
SMILES string
CC(=O)OI1(OC(C)=O)(OC(C)=O)OC(=O)c2ccccc12
InChI
1S/C13H13IO8/c1-8(15)19-14(20-9(2)16,21-10(3)17)12-7-5-4-6-11(12)13(18)22-14/h4-7H,1-3H3
InChI key
NKLCNNUWBJBICK-UHFFFAOYSA-N
Application
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Central nervous system
Supplementary Hazards
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service