Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

558346

Sigma-Aldrich

4-Methoxybenzhydrazide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H3COC6H4CONHNH2
CAS Number:
Molecular Weight:
166.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

136-140 °C (lit.)

functional group

amine
hydrazine

SMILES string

COc1ccc(cc1)C(=O)NN

InChI

1S/C8H10N2O2/c1-12-7-4-2-6(3-5-7)8(11)10-9/h2-5H,9H2,1H3,(H,10,11)

InChI key

REKQLYUAUXYJSZ-UHFFFAOYSA-N

Application

4-Methoxybenzhydrazide may be used to synthesize:
  • N′-[(E)-5-bromo-2-hydroxy-3-methoxybenzylidene]-4-methoxybenzohydrazide monohydrate
  • 2,5-bis(4-methoxyphenyl)-1,3,4-oxadiazole
  • hydrazone ligands, which readily forms ruthenium(II) hydrazone complexes via reaction with [RuHCl(CO)(PPh3)3]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

2, 5-Bis (4-methoxyphenyl)-1, 3, 4-oxadiazole.
Fun HK, et al.
Acta Crystallographica Section B, Structural Science, Crystal Engineering and Materials, 66(12), o3072-o3072 (2010)
Ruthenium (II) complexes containing 4-methoxybenzhydrazone ligands: synthesis, characterization, DNA binding, DNA cleavage, radical scavenging and in vitro cytotoxic activity.
Mohanraj M, et al.
Applied Organometallic Chemistry, 30(7), 550-560 (2016)
N'-[(E)-5-Bromo-2-hydroxy-3-methoxybenzylidene]-4-methoxybenzohydrazide monohydrate.
Reshma PR, et al.
Acta Crystallographica Section B, Structural Science, Crystal Engineering and Materials, 68(10), o2821-o2822 (2012)
Cheuk-Fai Chow et al.
Chemistry, an Asian journal, 3(8-9), 1324-1335 (2008-07-30)
Nine neutral dynamic metallosupramolecular polymers (metallodynamers) based on acyl hydrazone derived metal coordination centers (Co(2+), Ni(2+), Zn(2+), and Cd(2+)) were generated through self-assembly polymerization. These are linear coordination polymers with specific optical and mechanical properties. Monomer selection was found to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service