Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

549495

Sigma-Aldrich

(R)-(+)-1,1-Diphenyl-2-aminopropane

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H17N
CAS Number:
Molecular Weight:
211.30
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

optical activity

[α]20/D +16°, c = 1 in chloroform

mp

77-80 °C (lit.)

functional group

amine
phenyl

SMILES string

C[C@@H](N)C(c1ccccc1)c2ccccc2

InChI

1S/C15H17N/c1-12(16)15(13-8-4-2-5-9-13)14-10-6-3-7-11-14/h2-12,15H,16H2,1H3/t12-/m1/s1

InChI key

XNKICCFGYSXSAI-GFCCVEGCSA-N

Application

(R)-(+)-1,1-Diphenyl-2-aminopropane can be used as a precursor for the preparation of trisubstituted dihydroisoquinoline derivatives via Bischler-Napieralski reaction.

Legal Information

Product of Onyx Scientific, U.K.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The asymmetric Bischler-Napieralski reaction: preparation of 1, 3, 4-trisubstituted 1, 2, 3, 4-tetrahydroisoquinolines
Nicoletti M, et al.
Journal of the Chemical Society. Perkin Transactions 1, 116-121 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service