Skip to Content
MilliporeSigma

Skip To

548006

cis-5-Norbornene-exo-2,3-dicarboxylic anhydride

95%

Synonym(s):

cis -Norbornene-exo -2,3-dicarboxylic anhydride, cis -5-Norbornene-exo -2,3-dicarboxylic anhydride, cis -5-Norbornene-exo -anhydride, exo -5-Norbornene-2,3-dicarboxylic anhydride, exo -cis -5-Norbornene-2,3-dicarboxylic anhydride, exo -Norbornene dicarboxylic anhydride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C9H8O3
CAS Number:
Molecular Weight:
164.16
NACRES:
NA.23
PubChem Substance ID:
UNSPSC Code:
12352300
EC Number:
220-384-5
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

95%

mp

140-145 °C (lit.)

SMILES string

O=C1OC(=O)[C@@H]2[C@@H]3C[C@@H](C=C3)[C@H]12

InChI

1S/C9H8O3/c10-8-6-4-1-2-5(3-4)7(6)9(11)12-8/h1-2,4-7H,3H2/t4-,5+,6+,7-

InChI key

KNDQHSIWLOJIGP-RNGGSSJXSA-N


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
2178082476348.20365
assay

95%

assay

95%

assay

99%

assay

-

Quality Level

100

Quality Level

200

Quality Level

100

Quality Level

200

mp

140-145 °C (lit.)

mp

75-78 °C (lit.)

mp

165-167 °C (lit.)

mp

30-33 °C


pictograms

Health hazardCorrosion

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Resp. Sens. 1 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters