Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

545198

Sigma-Aldrich

Cucurbit[5]uril hydrate

contains acid of crystalization

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C30H30N20O10 · xH2O · xKCl · xHCl
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

solid

Quality Level

optical activity

[α]20/D −24°, c = 1 in H2O

contains

acid of crystalization

solubility

aqueous acid: soluble
saline: soluble
sugar solution: soluble

SMILES string

O.Cl.[Cl-].[K+].O=C1N2CN3C4C5N(CN6C7C8N(CN9C%10C%11N(CN%12C%13C%14N(CN1C%15C2N%16CN4C(=O)N5CN7C(=O)N8CN%10C(=O)N%11CN%13C(=O)N%14CN%15C%16=O)C%12=O)C9=O)C6=O)C3=O

InChI

1S/C30H30N20O10.2ClH.K.H2O/c51-21-31-1-32-12-14-36(22(32)52)4-40-16-18-44(26(40)56)8-48-20-19-47(29(48)59)7-43-17-15-39(25(43)55)3-35(21)13-11(31)33-2-34(12)24(54)38(14)6-42(16)28(58)46(18)10-50(20)30(60)49(19)9-45(17)27(57)41(15)5-37(13)23(33)53;;;;/h11-20H,1-10H2;2*1H;;1H2/q;;;+1;/p-1/t11-,12+,13+,14-,15-,16+,17+,18-,19-,20+;;;;

InChI key

VIWLZIOPOSWHFW-DVJLYPCZSA-M

Application

Host material for supramolecular chemistry.

Features and Benefits

Biologically inert; biocompatible

Other Notes

May contain ca.20% water. Contains acid of crystallization.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service