Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

540013

Sigma-Aldrich

(R)-(+)-Propylene carbonate

98%

Synonym(s):

(R)-1,2-Propanediol cyclic carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6O3
CAS Number:
Molecular Weight:
102.09
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

0.59 psi ( 20 °C)

Quality Level

Assay

98%

optical purity

ee: 95% (GLC)

refractive index

n20/D 1.422 (lit.)

bp

240 °C (lit.)

density

1.189 g/mL at 25 °C (lit.)

functional group

carbonate

SMILES string

C[C@@H]1COC(=O)O1

InChI

1S/C4H6O3/c1-3-2-6-4(5)7-3/h3H,2H2,1H3/t3-/m1/s1

InChI key

RUOJZAUFBMNUDX-GSVOUGTGSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Manufactured under license by Sterling Pharma Solutions Limited, using Jacobsen HKR technology.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

269.6 °F - closed cup

Flash Point(C)

132 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Steen H Matthiesen et al.
PloS one, 7(7), e40675-e40675 (2012-08-23)
Formamide is the preferred solvent to lower the melting point and annealing temperature of nucleic acid strands in in situ hybridization (ISH). A key benefit of formamide is better preservation of morphology due to a lower incubation temperature. However, in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service