Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

532738

Sigma-Aldrich

3-Fluorobenzenesulfonyl chloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4SO2Cl
CAS Number:
Molecular Weight:
194.61
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

refractive index

n20/D 1.529 (lit.)

bp

231-232 °C (lit.)

density

1.463 g/mL at 25 °C (lit.)

functional group

fluoro

SMILES string

Fc1cccc(c1)S(Cl)(=O)=O

InChI

1S/C6H4ClFO2S/c7-11(9,10)6-3-1-2-5(8)4-6/h1-4H

InChI key

OKYSUJVCDXZGKE-UHFFFAOYSA-N

General description

3-Fluorobenzenesulfonyl chloride, also known as m-fluorobenzenesulfonyl chloride, is a fluorinated arylsulfonyl chloride. It can be prepared from 1,3-benzenedisulfonyl fluoride.

Application

3-Fluorobenzenesulfonyl chloride may be used in the preparation of 1-(2-bromobenzyl)-2-(3-fluorophenyl)pyrrole and 1-(2-bromobenzyl)-2,5-bis(3-fluorophenyl)pyrrole.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Potassium fluoride catalyzed fluorodesulfonylations of aryl sulfonyl fluorides.
Van der Puy M.
The Journal of Organic Chemistry, 53(18), 4398-4401 (1988)
Palladium-Catalyzed Iterative C- H Bond Arylations: Synthesis of Medium-Size Heterocycles with a Bridgehead Nitrogen Atom.
Hagui W, et al.
ChemCatChem, 7(21), 3544-3554 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service