Skip to Content
MilliporeSigma
All Photos(1)

Documents

531944

Sigma-Aldrich

(R,R)-N,N′-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminochromium(III) chloride

Synonym(s):

(-)-Chloro[[2,2′-[(1R,2R)-1,2-cyclohexanediylbis[(nitrilo-κN)methylidyne]]bis[4,6-bis(1,1-dimethylethyl)phenolato-κO]](2-)]chromium, (SP-5-13)-Chloro[[2,2′-[(1R,2R)-1,2-cyclohexanediylbis[(nitrilo-κN)methylidyne]]bis[4,6-bis(1,1-dimethylethyl)phenolato-κO]](2-)]chromium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H52ClCrN2O2
CAS Number:
Molecular Weight:
632.26
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

mp

>350 °C (lit.)

SMILES string

CC(C)(C)c1cc2\C=N\[C@@H]3CCCC[C@H]3\N=C\c4cc(cc(c4O[Cr](Cl)Oc2c(c1)C(C)(C)C)C(C)(C)C)C(C)(C)C

InChI

1S/C36H54N2O2.ClH.Cr/c1-33(2,3)25-17-23(31(39)27(19-25)35(7,8)9)21-37-29-15-13-14-16-30(29)38-22-24-18-26(34(4,5)6)20-28(32(24)40)36(10,11)12;;/h17-22,29-30,39-40H,13-16H2,1-12H3;1H;/q;;+3/p-3/b37-21+,38-22+;;/t29-,30-;;/m1../s1

InChI key

ZWANLSXDTHQDNO-OHRASPNLSA-K

Application

Catalyst used for:
  • Asymmetric anti-Mannich reactions
  • Coupling reactions
  • Ring-opening and Diels-Alder reactions
  • Allylic C-H oxidation of terminal alkenes

Legal Information

Sold under license from Sterling Pharma Solutions Limited.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service