Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

531316

Sigma-Aldrich

N-(tert-Butoxycarbonyl)-2-bromoaniline

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4HNCO2C(CH3)3
CAS Number:
Molecular Weight:
272.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.54 (lit.)

bp

110 °C/0.3 mmHg (lit.)

density

1.342 g/mL at 25 °C (lit.)

functional group

amine
bromo

SMILES string

CC(C)(C)OC(=O)Nc1ccccc1Br

InChI

1S/C11H14BrNO2/c1-11(2,3)15-10(14)13-9-7-5-4-6-8(9)12/h4-7H,1-3H3,(H,13,14)

InChI key

UQBGKDLSIIHUEZ-UHFFFAOYSA-N

General description

N-(tert-Butoxycarbonyl)-2-bromoaniline, also known as tert-butyl-N-(2-bromophenyl)carbamate, is an N-Boc-protected o-bromoaniline.1 It reacts with ethyl perfluorooctanoate in the presence of tert-butyllithium to form the corresponding 1-hydroxy-1H-perfluorooctyl compound.

Application

N-(tert-Butoxycarbonyl)-2-bromoaniline may be used in the synthesis of N-boc-protected o-aryl anilines, which undergoes coupling reaction with bromoacetylenes to form the corresponding arene?ynamides.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yousuke Yamaoka et al.
The Journal of organic chemistry, 80(2), 957-964 (2014-12-30)
A Brønsted acid-promoted arene-ynamide cyclization has been developed to construct the 3H-pyrrolo[2,3-c]quinolines. This reaction consists of the generation of a highly reactive keteniminium intermediate from arene-ynamide activated by a Brønsted acid and electrophilic aromatic substitution reaction to give arene-fused quinolines
Novel reduction of perfluoroalkyl ketones with lithium alkoxides.
Sokeirik YS, et al.
Journal of Fluorine Chemistry, 127(1), 150-152 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service