Skip to Content
MilliporeSigma
All Photos(1)

Documents

531219

Sigma-Aldrich

Bromoform-13C

≥99 atom % 13C, ≥99% (CP), contains copper as stabilizer

Synonym(s):

Tribromomethane-13C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13CHBr3
CAS Number:
Molecular Weight:
253.72
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

≥99 atom % 13C

Assay

≥99% (CP)

contains

copper as stabilizer

refractive index

n20/D 1.596 (lit.)

bp

146-150 °C (lit.)

mp

5-8 °C (lit.)

mass shift

M+1

SMILES string

Br[13CH](Br)Br

InChI

1S/CHBr3/c2-1(3)4/h1H/i1+1

InChI key

DIKBFYAXUHHXCS-OUBTZVSYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service