Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

526320

Sigma-Aldrich

Polycaprolactone-block-polytetrahydrofuran-block-polycaprolactone

Synonym(s):

Poly THF, Polymeg, Polytetramethylene ether glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO[(CH2)5CO2]x [(CH2)4O]y[CO(CH2)5O]zH
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

mol wt

average Mn ~2,000

mp

30-33 °C (lit.)

density

1.05 g/mL at 25 °C (lit.)

Mw/Mn

1.3

SMILES string

OCCCCO

InChI

1S/C4H10O2/c5-3-1-2-4-6/h5-6H,1-4H2

InChI key

WERYXYBDKMZEQL-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service