Skip to Content
MilliporeSigma
All Photos(1)

Documents

522902

Sigma-Aldrich

(S)-(+)-2-Methylpiperidine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13N
CAS Number:
Molecular Weight:
99.17
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

optical activity

[α]20/D +35°, c = 3.5 in hexane

refractive index

n20/D 1.446 (lit.)

bp

117-121 °C (lit.)

density

0.823 g/mL at 25 °C (lit.)

SMILES string

C[C@H]1CCCCN1

InChI

1S/C6H13N/c1-6-4-2-3-5-7-6/h6-7H,2-5H2,1H3/t6-/m0/s1

InChI key

NNWUEBIEOFQMSS-LURJTMIESA-N

Application

(S)-(+)-2-Methylpiperidine has been used in the preparation of (S)-N-(3-(2-pipecolin-1-yl)propyl)phthalimide by reacting with N-(3-bromopropyl)phthalimide. It may also be used as a starting material in the multi-step synthesis of (2S,6S)-(+)-solenopsin A.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

60.0 °F - closed cup

Flash Point(C)

15.56 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anodic Cyanation of (-)-N-Phenyl-2-Methylpiperidine: A Short Synthesis of (+)-Solenopsin A and (+)-Isosolenopsin A.
Girard N, et al.
Synthetic Communications, 35(5), 711-723 (2005)
Noncyclam Tetraamines Inhibit CXC Chemokine Receptor Type 4 and Target Glioma-Initiating Cells.
Ros-Blanco L, et al.
Journal of Medicinal Chemistry, 55(17), 7560-7570 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service