Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

520365

Sigma-Aldrich

Copper-tin alloy

spherical powder, −200 mesh

Synonym(s):

Bronze, Sn5Cu84

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Sn5Cu84
CAS Number:
MDL number:
UNSPSC Code:
31152304
PubChem Substance ID:
NACRES:
NA.23

form

spherical powder

Quality Level

composition

copper, 90 wt. %
Tin, 10 wt. %

particle size

−200 mesh

SMILES string

[Cu].[Sn]

InChI

1S/Cu.Sn

InChI key

KUNSUQLRTQLHQQ-UHFFFAOYSA-N

Related Categories

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Mechanical alloying is a “brute force” method of affecting alloying and chemical reactions. The mixture of reactant powders and several balls are placed in the milling jar of a high-energy ball mill, for example, a shaker mill or a planetary mill.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service