Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

510750

Sigma-Aldrich

1-Chloro-4-(phenylethynyl)benzene

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4C≡CC6H5
CAS Number:
Molecular Weight:
212.67
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

81-83 °C (lit.)

SMILES string

Clc1ccc(cc1)C#Cc2ccccc2

InChI

1S/C14H9Cl/c15-14-10-8-13(9-11-14)7-6-12-4-2-1-3-5-12/h1-5,8-11H

InChI key

YMZAAGCDWVIPNL-UHFFFAOYSA-N

General description

1-Chloro-4-(phenylethynyl)benzene is an internal alkyne that can be prepared by palladium-catalyzed Kumada cross-coupling reaction.

Application

1-Chloro-4-(phenylethynyl)benzene may be used in the preparation of 1-(4-chlorophenyl)-4-methyl-2-phenylnaphthalene and 2-(4-chlorophenyl)-4-methyl-1-phenylnaphthalene as a 1:1 mixture.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of Internal Alkynes by Pd(PPh3)4/TMEDA-Catalyzed Kumada Cross-Coupling of Alkynyl Halides with Grignard Reagents.
Zhang MM, et al.
European Journal of Organic Chemistry, 2014(30), 6769-6773 (2014)
Practical synthesis of polysubstituted naphthalene derivatives via HNTf2-catalyzed benzannulation reaction.
Ponra S, et al.
ARKIVOC (Gainesville, FL, United States), 2, 62-81 (2016)
Alisa Bahar Beydogan et al.
The Journal of pharmacy and pharmacology, 71(3), 408-416 (2018-11-15)
A large amount of fructose is metabolized in the liver and causes hepatic functional damage. Δ9 -tetrahydrocannabinol (THC) is known as a therapeutic agent for clinical and experimental applications. The study aims to investigate the effects of THC treatment on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service