Skip to Content
MilliporeSigma
All Photos(1)

Documents

50790

Sigma-Aldrich

Gold(III) chloride hydrate

~50% Au basis

Synonym(s):

Auric chloride, Chloroauric acid, Hydrogen tetrachloroaurate(III) hydrate, Tetrachloroauric(III) acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HAuCl4 · aq
CAS Number:
Molecular Weight:
339.79 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Quality Level

form

powder or crystals

concentration

~50% Au

color

yellow

density

3.9 g/mL at 25 °C

SMILES string

O.Cl.Cl[Au](Cl)Cl

InChI

1S/Au.4ClH.H2O/h;4*1H;1H2/q+3;;;;;/p-3

InChI key

OVJUDSKPNIBLOO-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

General description

Gold (III) chloride hydrate,also known as chloroauric acid, is used as precursor to synthesize gold nanoparticles. It also exihibits very high catalytic activity.

Application

Used as a catalyst in asymmetric aldol reactions, diboration of vinylarenes, dehydrogenative dimerizations of trialkystannanes, carbonylation of olefins and amines, C-C, C-O bond couplings, oxidation of sulfides, selective oxidation of thioethers.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT RE 2 Oral

Target Organs

Kidney

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The crystal structure of gold (III) chloride
Acta Crystallographica, 11(4), 248-288 (1958)
Raquel Casado et al.
Journal of the American Chemical Society, 125(39), 11925-11935 (2003-09-25)
Different inorganic and organometallic gold(III) and gold(I) complexes have been tested in the addition of water and methanol to terminal alkynes. Anionic and neutral organometallic gold(III) compounds can efficiently mediate these reactions in neutral media in refluxing methanol. The compounds

Articles

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service