Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

489662

Sigma-Aldrich

Palmitic acid-2,2-d2

98 atom % D

Synonym(s):

Hexadecanoic-2,2-d2 acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)13CD2CO2H
CAS Number:
Molecular Weight:
258.44
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % D

Assay

99% (CP)

form

solid

mp

61-64 °C

mass shift

M+2

SMILES string

[2H]C([2H])(CCCCCCCCCCCCCC)C(O)=O

InChI

1S/C16H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16(17)18/h2-15H2,1H3,(H,17,18)/i15D2

InChI key

IPCSVZSSVZVIGE-DOBBINOXSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kristina Glenske et al.
Bone reports, 13, 100707-100707 (2020-09-12)
The long-term effects of palmitate (PA) on osteogenic differentiation capacity of human mesenchymal stromal cells (hMSCs) were investigated by cultivating the cells in osteogenic differentiation medium (O-w/o) and osteogenic medium containing PA (O-BSA-PA) for 21 days. Osteogenic medium containing BSA (O-BSA)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service