Skip to Content
MilliporeSigma
All Photos(1)

Documents

489522

Sigma-Aldrich

Glycine-13C2,15N

98 atom % 15N, 99 atom % 13C

Synonym(s):

13C and 15N Labeled glycine, Aminoacetic acid-13C2,15N, Aminoethanoic acid-13C2,15N

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H215N13CH213CO2H
CAS Number:
Molecular Weight:
78.05
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

99 atom % 13C
98 atom % 15N

Quality Level

Assay

99% (CP)

form

solid

technique(s)

bio NMR: suitable

mp

240 °C (dec.) (lit.)

mass shift

M+3

SMILES string

[15NH2][13CH2][13C](O)=O

InChI

1S/C2H5NO2/c3-1-2(4)5/h1,3H2,(H,4,5)/i1+1,2+1,3+1

InChI key

DHMQDGOQFOQNFH-VMIGTVKRSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna-Sara Claeson et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1104, 67-72 (2018-11-18)
Variability in the levels of GSH and GSSG in plasma is suggested to derive from inadequate pre-processing methods. The aim of this study was to develop a protocol for comparable and reliable measurements of GSH/GSSG. Venous blood from 8 healthy
Eunus S Ali et al.
Molecular cell, 78(6), 1178-1191 (2020-06-03)
The RAS-ERK/MAPK (RAS-extracellular signal-regulated kinase/mitogen-activated protein kinase) pathway integrates growth-promoting signals to stimulate cell growth and proliferation, at least in part, through alterations in metabolic gene expression. However, examples of direct and rapid regulation of the metabolic pathways by the
Curtis W Bacon et al.
Molecular cell, 78(6), 1133-1151 (2020-05-14)
Precise control of the RNA polymerase II (RNA Pol II) cycle, including pausing and pause release, maintains transcriptional homeostasis and organismal functions. Despite previous work to understand individual transcription steps, we reveal a mechanism that integrates RNA Pol II cycle

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service