489409
Fluoromethane-13C
99 atom % 13C
Synonym(s):
Methyl-13C fluoride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
13CH3F
CAS Number:
Molecular Weight:
35.03
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.12
Recommended Products
vapor density
1.231 (vs air)
Quality Level
vapor pressure
44.2 atm ( 21.1 °C)
isotopic purity
99 atom % 13C
bp
−77.9 °C (lit.)
mp
−141.8 °C (lit.)
mass shift
M+1
SMILES string
[13CH3]F
InChI
1S/CH3F/c1-2/h1H3/i1+1
InChI key
NBVXSUQYWXRMNV-OUBTZVSYSA-N
Related Categories
Packaging
100 mL quantity is packaged in a 25 mL Sure/Pac™ cylinder with brass 1/4" male NPT valve. Nominal gas pressure at 21ºC is 32 psig for 100 mL. This pressure is slightly above atmospheric pressure. Care must be taken when extracting this product from the cylinder.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Recommended products
Brass hose adapter Z146811 or brass body mini gas regulator Z513539 is recommended.
Legal Information
Sure/Pac is a trademark of Sigma-Aldrich Co. LLC
hose barb
Product No.
Description
Pricing
regulator
Product No.
Description
Pricing
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Gas 1 - Press. Gas Liquefied gas
Storage Class Code
2A - Gases
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Lingchun Song et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 12(28), 7458-7466 (2006-07-29)
The recently developed (L. Song, W. Wu, Q. Zhang, S. Shaik, J. Phys. Chem. A 2004, 108, 6017) valence bond method coupled with a polarized continuum model (VBPCM) has been applied to the identity SN2 reaction of halides in the
Patrice L Jackson et al.
European journal of medicinal chemistry, 51, 42-51 (2012-03-16)
The x-ray crystal structure of 3-((5-methylisoxazol-3-yl)amino)-5-methylcyclohex-2-enone (12b) and 3-((5-methylisoxazolyl-3-yl)amino)-5,5-dimethylcyclohex-2-enone (12c) were determined and correlated to their anticonvulsant activity in mice and rats. A hypothesis for the toxicity of the analogs are advanced. In addition, a series of 5-methyl-N-(3-oxocyclohex-1-enyl)-isoxazole-3-carboxamides were synthesized
Xiang Zhao et al.
The journal of physical chemistry. A, 110(36), 10607-10618 (2006-09-08)
Reactions of CH(3)F have been surveyed systematically at room temperature with 46 different atomic cations using an inductively coupled plasma/selected-ion flow tube tandem mass spectrometer. Rate coefficients and product distributions were measured for the reactions of fourth-period atomic ions from
Jos Oomens et al.
The journal of physical chemistry. A, 112(43), 10774-10783 (2008-10-04)
The multiple CF bond character of PhCFMe (+) ions has been examined by means of theory, vibrational spectroscopy of the gaseous ions, and unimolecular decomposition chemistry. Atoms in Molecules analysis of DFT wave functions gives a CF bond order of
Robert E Rosenberg
The journal of physical chemistry. A, 116(44), 10842-10849 (2012-10-13)
Fluorinated organic compounds have become increasingly important in the pharmaceutical and agricultural industries. However, even the simplest aspects of these compounds are still not well understood. For instance, it is an open question as to whether fluoroorganics can form a
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service