Skip to Content
MilliporeSigma

488232

Bromobenzene-13C6

99 atom % 13C

Synonym(s):

1-bromo-[1,2,3,4,5,6-13C6]benzene

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
13C6H5Br
CAS Number:
Molecular Weight:
162.96
NACRES:
NA.12
PubChem Substance ID:
UNSPSC Code:
12352101
MDL number:
Isotopic purity:
99 atom % 13C
Assay:
99% (CP)
Mass shift:
M+6
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


isotopic purity

99 atom % 13C

Quality Segment

assay

99% (CP)

technique(s)

protein expression: suitable

refractive index

n20/D 1.559 (lit.)

bp

156 °C (lit.)

mp

−31 °C (lit.)

density

1.547 g/mL at 25 °C

mass shift

M+6

SMILES string

Br[13c]1[13cH][13cH][13cH][13cH][13cH]1

InChI

1S/C6H5Br/c7-6-4-2-1-3-5-6/h1-5H/i1+1,2+1,3+1,4+1,5+1,6+1

InChI key

QARVLSVVCXYDNA-IDEBNGHGSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.


signalword

Warning

Hazard Classifications

Aquatic Chronic 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

123.8 °F - closed cup

flash_point_c

51 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library