Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

487783

Sigma-Aldrich

Acetanilide-15N

98 atom % 15N

Synonym(s):

N-Phenylacetamide-15N

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO15NHC6H5
CAS Number:
Molecular Weight:
136.16
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

98 atom % 15N

Quality Level

Assay

99% (CP)

form

solid

bp

304 °C (lit.)

mp

113-115 °C (lit.)

density

1.230 g/mL at 25 °C

mass shift

M+1

SMILES string

CC(=O)[15NH]c1ccccc1

InChI

1S/C8H9NO/c1-7(10)9-8-5-3-2-4-6-8/h2-6H,1H3,(H,9,10)/i9+1

InChI key

FZERHIULMFGESH-QBZHADDCSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

321.8 °F - closed cup

Flash Point(C)

161.00 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service