Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

484369

Sigma-Aldrich

Cyclopentadienylhafnium(IV) trichloride

97%

Synonym(s):

Trichloro(η5-2,4-cyclopentadien-1-yl) hafnium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5Cl3Hf
CAS Number:
Molecular Weight:
349.94
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

reaction suitability

core: hafnium
reagent type: catalyst

impurities

5% benzene

mp

125 °C (dec.) (lit.)

SMILES string

Cl[Hf](Cl)Cl.[CH]1[CH][CH][CH][CH]1

InChI

1S/C5H5.3ClH.Hf/c1-2-4-5-3-1;;;;/h1-5H;3*1H;/q;;;;+3/p-3

InChI key

OAEWLULEWYPRGF-UHFFFAOYSA-K

Application

Reactant for synthesis of hafnocene dihalides

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1A - Eye Dam. 1 - Muta. 1B - Skin Corr. 1B - STOT RE 2

Target Organs

Blood

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service