Skip to Content
MilliporeSigma
All Photos(1)

Documents

483397

Sigma-Aldrich

2-Bromoethyl isocyanate

≥90%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCH2CH2NCO
CAS Number:
Molecular Weight:
149.97
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥90%

refractive index

n20/D 1.483 (lit.)

bp

158 °C (lit.)

density

1.66 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

BrCCN=C=O

InChI

1S/C3H4BrNO/c4-1-2-5-3-6/h1-2H2

InChI key

JLHTVZLEHOQZBM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.0 °F - closed cup

Flash Point(C)

112.8 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michaël Bosco et al.
Methods (San Diego, Calif.), 180, 69-78 (2020-06-09)
We present herein the synthesis of biotin-functionalized polymers (BNAPols) that have been developed for the fixation of membrane proteins (MPs) onto surfaces. BNAPols were synthesized by free-radical polymerization of a tris(hydroxymethyl)acrylamidomethane (THAM)-derived amphiphilic monomer in the presence of a thiol-based
Datong Wu et al.
The Analyst, 145(9), 3395-3400 (2020-04-03)
Here, ionized chiral carbon dots, (S,S)-C-dots-1 (λex = 430 nm, λem = 480 nm), were synthesized via a facile route with relatively high quantum yield (∼24.4%) and used as a fluorescent chiral sensor. One of the advantages of the synthetic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service