Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

482331

Sigma-Aldrich

4-Chlorobutyl ether

98%

Synonym(s):

Bis(4-chlorobutyl) ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[Cl(CH2)4]2O
CAS Number:
Molecular Weight:
199.12
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.459 (lit.)

bp

129-131 °C/10 mmHg (lit.)

density

1.081 g/mL at 25 °C (lit.)

functional group

chloro
ether

SMILES string

ClCCCCOCCCCCl

InChI

1S/C8H16Cl2O/c9-5-1-3-7-11-8-4-2-6-10/h1-8H2

InChI key

PVBMXMKIKMJQRK-UHFFFAOYSA-N

Related Categories

Application

4-Chlorobutyl ether is generally used as an ether-containing linker while synthesizing organic compounds.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fabrication of surface-modified hydrogels with polyion complex for controlled release.
Takemoto Y, et al.
Chemistry of Materials, 22(9), 2923-2929 (2010)
Single-molecule conductance determinations on HS (CH2) 4O (CH2) 4SH and HS (CH2) 2O (CH2) 2O (CH2) 2SH, and comparison with alkanedithiols of the same length.
Scullion LE, et al.
Journal of Physics. Condensed Matter : An Institute of Physics Journal, 24(16), 164211-164211 (2012)
High-dilution cyclization of polyoxapentacosanodinitriles.
Newman MS, et al.
The Journal of Organic Chemistry, 40(20), 2863-2870 (1975)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service