Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

481963

Sigma-Aldrich

Poly(dimethylsiloxane), hydroxy terminated

viscosity ~750 cSt 

Synonym(s):

PDMS

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

refractive index

n20/D 1.406

viscosity

~750 cSt

density

0.97 g/mL at 25 °C

SMILES string

C[Si](C)(O)O

InChI key

XCLIHDJZGPCUBT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Fabrication of silicone modified polyurethane matrix superhydrophobic coating with hydroxy-terminated polydimethylsiloxane modified SiO2 nanoparticles: This study uses hydroxy-terminated PDMS as a modifier to hydrophobically modify SiO2 nanoparticles (Y Cui, Y Li, D Huan, D Zeng, Y Yang, 2022).

  • High glass transition temperature shape‐memory materials: Hydroxyl‐terminated polydimethylsiloxane‐modified cyanate ester: Hydroxyl-terminated PDMS is used to modify cyanate ester, enhancing shape-memory properties (Z Li, J Hu, L Ma, H Liu, 2020).

  • The preparation and properties study of polydimethylsiloxane-based coatings modified by epoxy resin: This research explores modifying hydroxyl-terminated PDMS with siliconized epoxy resin for improved coating properties (C Zhou, R Li, W Luo, Y Chen, H Zou, M Liang, 2016).

  • Synthesis and structure of calcium silicate hydrate (CSH) modified by hydroxyl-terminated polydimethylsiloxane (PDMS): Hydroxyl-terminated PDMS is used to modify CSH, enhancing its properties for construction applications (Z Zhu, Z Wang, Y Zhou, Y Wei, A She, 2021).

  • Crosslinked polymer networks of poly (ethylene glycol) (PEG) and hydroxyl terminated poly (dimethyl siloxane) (HTPDMS) as polymeric phase change material: This study develops PEG and hydroxyl-terminated PDMS crosslinked networks for phase change materials (S Sundararajan, AB Samui, PS Kulkarni, 2019).

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.

Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.

Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.

Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.

Related Content

We offer PDMS surface modification kit which allows for durable hydrophilization of various polydimethylsiloxane (PDMS) surfaces.

We offer PDMS surface modification kit which allows for durable hydrophilization of various polydimethylsiloxane (PDMS) surfaces.

We offer PDMS surface modification kit which allows for durable hydrophilization of various polydimethylsiloxane (PDMS) surfaces.

We offer PDMS surface modification kit which allows for durable hydrophilization of various polydimethylsiloxane (PDMS) surfaces.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service