Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

476307

Sigma-Aldrich

3,4′-Oxydianiline

97%

Synonym(s):

3,4′-DAPE, 3,4′-Diaminodiphenyl ether, 3,4′-Oxydiphenylamine, 3-(4-Aminophenoxy)aniline, 3-(4-Aminophenoxy)benzenamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O(C6H4NH2)2
CAS Number:
Molecular Weight:
200.24
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

solid

bp

207 °C/1 mmHg (lit.)

mp

67-71 °C (lit.)

SMILES string

Nc1ccc(Oc2cccc(N)c2)cc1

InChI

1S/C12H12N2O/c13-9-4-6-11(7-5-9)15-12-3-1-2-10(14)8-12/h1-8H,13-14H2

InChI key

ZBMISJGHVWNWTE-UHFFFAOYSA-N

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Muta. 2 - Repr. 2 - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service