Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

475424

Sigma-Aldrich

PSS-Octavinyl substituted

Synonym(s):

1,3,5,7,9,11,13,15-Octavinylpentacyclo[9.5.1.13,9.15,15.17,13]octasiloxane, Octavinyloctasilasesquioxane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H24O12Si8
CAS Number:
Molecular Weight:
633.04
Beilstein:
2320666
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

solid

Quality Level

mp

>350 °C (lit.)

SMILES string

C=C[Si]12O[Si]3(O[Si]4(O[Si](O1)(O[Si]5(O[Si](O2)(O[Si](O3)(O[Si](O4)(O5)C=C)C=C)C=C)C=C)C=C)C=C)C=C

InChI

1S/C16H24O12Si8/c1-9-29-17-30(10-2)20-33(13-5)22-31(11-3,18-29)24-35(15-7)25-32(12-4,19-29)23-34(14-6,21-30)27-36(16-8,26-33)28-35/h9-16H,1-8H2

InChI key

ZWCNRMDCQDJIRL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xianfeng Zhou et al.
Biomaterials, 50, 1-9 (2015-03-05)
Inspired by nature's toughening mechanisms, we designed a new polyhedral oligomeric silsesquioxane (POSS)-derived hybrid glass (PHG) that has covalent interactions on the molecular scale between the inorganic POSS cage and organic phase. These features allow "elastic deformation" of the inorganic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service