Skip to Content
MilliporeSigma
All Photos(1)

Documents

469947

Sigma-Aldrich

Tris(dimethylamido)aluminum(III)

Synonym(s):

Hexakis(dimethylamido)dialuminum, Tris(dimethylamino)alane dimer

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Al(N(CH3)2)3
CAS Number:
Molecular Weight:
159.21
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

solid

Quality Level

reaction suitability

core: aluminum

mp

82-84 °C (lit.)

density

0.865 g/mL at 25 °C (lit.)

SMILES string

CN(C)[Al](N(C)C)N(C)C.CN(C)[Al](N(C)C)N(C)C

InChI

1S/6C2H6N.2Al/c6*1-3-2;;/h6*1-2H3;;/q6*-1;2*+3

InChI key

JGZUJELGSMSOID-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Atomic number of base material: 13 Aluminum

Application

Tris(dimethylamido)aluminum(III) may be used as a precursor to aluminum nitride (AlN) thin films by organometallic chemical vapor deposition (OMCVD).

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B - Water-react 1

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

70.0 °F - closed cup

Flash Point(C)

21.1 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tris (dimethylamido) aluminum (III): An overlooked atomic layer deposition precursor.
Buttera SC, et al.
Journal of Vacuum Science & Technology. A, Vacuum, Surfaces, And Films, 35(1), 01B128-01B128 (2017)

Articles

Thin film photovoltaic devices have become increasingly important in efficiently harnessing solar energy to meet consumer demand.

Nanomaterials are considered a route to the innovations required for large-scale implementation of renewable energy technologies in society to make our life sustainable.

Nanomaterials are considered a route to the innovations required for large-scale implementation of renewable energy technologies in society to make our life sustainable.

Nanomaterials are considered a route to the innovations required for large-scale implementation of renewable energy technologies in society to make our life sustainable.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service