Skip to Content
MilliporeSigma
All Photos(1)

Documents

468037

Sigma-Aldrich

cis-1,4-Diacetoxy-2-butene

95%

Synonym(s):

cis-2-Butene-1,4-diol diacetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2CH2CH=CHCH2O2CCH3
CAS Number:
Molecular Weight:
172.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

refractive index

n20/D 1.443 (lit.)

bp

120-121 °C/18 mmHg (lit.)

density

1.08 g/mL at 25 °C (lit.)

SMILES string

[H]\C(COC(C)=O)=C(/[H])COC(C)=O

InChI

1S/C8H12O4/c1-7(9)11-5-3-4-6-12-8(2)10/h3-4H,5-6H2,1-2H3/b4-3-

InChI key

VZUAUHWZIKOMFC-ARJAWSKDSA-N

General description

cis-1,4-Diacetoxy-2-butene is an ester. Ruthenium olefin metathesis catalysts having cyclic (alkyl)(amino)carbenes effectively catalyzes the cross-metathesis of cis-1,4-diacetoxy-2-butene with allylbenzene. cis-1,4-Diacetoxy-2-butene on pyrolysis affords 1-acetoxy-1,3-butadiene, a diacetate and an isomeric 1,2-diacetoxy-3-butene. Mechanism of pyrolysis has been investigated.

Application

cis-1,4-Diacetoxy-2-butene may be used in the preparation of hindered Diels-Alder adducts. It may be used for the synthesis of 1,2,3,4-tetrahydro-2-vinylquinoxalines, via Palladium-catalyzed tandem allylation of 1,2-phenylenediamines.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pyrolysis of Esters. V. Mechanism of 1, 4-Elimination1.
Bailey WJ and BARCLAY Jr R.
The Journal of Organic Chemistry, 21(3), 328-331 (1956)
Palladium-catalyzed tandem allylation of 1, 2-phenylenediamines with cis-1, 4-diacetoxy-2-butene.
Yang SC, et al.
Tetrahedron Letters, 45(25), 4951-4954 (2004)
Donde R Anderson et al.
Organometallics, 27(4), 563-566 (2008-06-28)
The evaluation of ruthenium olefin metathesis catalysts 4-6 bearing cyclic (alkyl)(amino)carbenes (CAACs) in the cross-metathesis of cis-1,4-diacetoxy-2-butene (7) with allylbenzene (8) and the ethenolysis of methyl oleate (11) is reported. Relative to most NHC-substituted complexes, CAAC-substituted catalysts exhibit lower E/Z
Kerstin Rosmarion et al.
Applied spectroscopy, 59(3), 366-375 (2005-05-20)
Hindered Diels-Alder adducts have been prepared from 3,6-dibromophencyclone, 2, with cis-1,4-diacetoxy-2-butene, 3; cis-2-butene-1,4-diol, 4; and N-benzylmaleimide, 5. The adduct from the parent phencyclone, 1, with N-benzylmaleimide was prepared for comparison. One- and two-dimensional (1D and 2D) proton and carbon-13 NMR

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service