Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

467456

Sigma-Aldrich

2-Methylthianaphthene

97%

Synonym(s):

2-Methylbenzo[b]thiophene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H8S
CAS Number:
Molecular Weight:
148.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

47-52 °C (lit.)

SMILES string

Cc1cc2ccccc2s1

InChI

1S/C9H8S/c1-7-6-8-4-2-3-5-9(8)10-7/h2-6H,1H3

InChI key

BLZKSRBAQDZAIX-UHFFFAOYSA-N

General description

2-Methylthianaphthene is a 2-substituted thianaphthene derivative. It can be synthesized by reacting 2-thianaphthenyllithium with methyl p-toluenesulfonate.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lei Wang et al.
Journal of hazardous materials, 342, 758-769 (2017-09-20)
The ZrO
Some 2-Substituted Thianaphthenes Derived from 2-Thianaphthenyllithium1.
Shirley DA and Cameron MD.
Journal of the American Chemical Society, 74(3), 664-665 (1952)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service