Skip to Content
MilliporeSigma
All Photos(1)

Documents

467367

Sigma-Aldrich

4-(3-Phenylpropyl)pyridine N-oxide

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H15NO
CAS Number:
Molecular Weight:
213.28
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

mp

58-65 °C (lit.)

SMILES string

[O-][n+]1ccc(CCCc2ccccc2)cc1

InChI

1S/C14H15NO/c16-15-11-9-14(10-12-15)8-4-7-13-5-2-1-3-6-13/h1-3,5-6,9-12H,4,7-8H2

InChI key

OOFBEJNEUVLZOW-UHFFFAOYSA-N

General description

4-(3-Phenylpropyl)pyridine N-oxide (P3NO, 4-PPPyNO) participates in catalytic media for manganese-salen complex in the asymmetric epoxidation of indene.

Application

4-(3-Phenylpropyl)pyridine N-oxide may be employed as a donor ligand in the managnese-salen complex catalyzed asymmetric epoxidation of indene and styrene.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The role of 4-(3-phenylpropyl) pyridine N-Oxide (P3NO) in the manganese-salen-catalyzed asymmetric epoxidation of indene.
Senanayake CH, et al.
Tetrahedron Letters, 37(19), 3271-3274 (1996)
Easily recyclable polymeric ionic liquid-functionalized chiral salen Mn (III) complex for enantioselective epoxidation of styrene.
Tan R, et al.
J. Catal., 263(2), 284-291 (2009)
David L. Hughes et al.
The Journal of organic chemistry, 62(7), 2222-2229 (1997-04-04)
The asymmetric epoxidation of indene using aqueous NaOCl, catalyzed by Jacobsen's chiral manganese salen complex, provides indene oxide in 90% yield and 85-88% enantioselectivity. The axial ligand, 4-(3-phenylpropyl)pyridine N-oxide (P(3)NO), increases the rate of epoxidation without affecting enantioselectivity and also
A novel polymeric chiral salen Mn (III) complex as solvent-regulated phase transfer catalyst in the asymmetric epoxidation of styrene.
Tan R, et al.
J. Mol. Catal. A: Chem., 259(1), 125-132 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service