Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

464813

Sigma-Aldrich

Tetraammineplatinum(II) hydroxide hydrate

Synonym(s):

Tetrammineplatinum dihydroxide hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pt(NH3)4(OH)2·xH2O
Molecular Weight:
297.21 (anhydrous basis)
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

core: platinum
reagent type: catalyst

mp

211 °C (dec.) (lit.)

SMILES string

[Pt].N.O

InChI

1S/4H3N.3H2O.Pt/h4*1H3;3*1H2;/q;;;;;;;+2/p-2

InChI key

YNZKWSKEPMJRMB-UHFFFAOYSA-L

General description

Tetraammineplatinum(II) hydroxide hydrate is an inorganic platinum catalyst generally used as a precursor for the preparation of platinum complexes.

Application

Tetraammineplatinum(II) hydroxide hydrate can be used as a platinum precursor to synthesze Pt/CeO2 catalysts which is used in water-gas shift reaction.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comparison of the effects of the catalyst preparation method and CeO 2 morphology on the catalytic activity of Pt/CeO 2 catalysts for the water-gas shift reaction
Lee YL, et al.
Catalysis Science & Technology, 10(18), 6299-6308 (2020)
Transformation of ethylbenzene-m-xylene feed over MCM-22 zeolites with different acidities
Mih'alyi MR, et al.
Applied Catalysis A: General, 476, 19-25 (2014)
Nanocrystalline Pt thin films prepared by electrostatic spray deposition for automotive exhaust gas treatment
Lintanf A, et al.
Solid State Ionics, 177(39-40), 3491-3499 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service