Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

464007

Sigma-Aldrich

(S)-(−)-3-Butyn-2-ol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HC≡CCH(OH)CH3
CAS Number:
Molecular Weight:
70.09
Beilstein:
4652597
MDL number:
UNSPSC Code:
12352003
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

optical activity

[α]22/D −45°, neat

refractive index

n20/D 1.426 (lit.)

bp

108-111 °C (lit.)

density

0.883 g/mL at 20 °C (lit.)

functional group

hydroxyl

SMILES string

C[C@H](O)C#C

InChI

1S/C4H6O/c1-3-4(2)5/h1,4-5H,2H3/t4-/m0/s1

InChI key

GKPOMITUDGXOSB-BYPYZUCNSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used in a ruthenium-catalyzed polymerization providing helical polyacetylenes.

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

78.8 °F - closed cup

Flash Point(C)

26 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pablo Morán-Poladura et al.
Beilstein journal of organic chemistry, 9, 2120-2128 (2013-11-10)
An efficient entry to the preparation of elusive 4-unsubstituted-3-iodo-2H-chromenes has been accomplished as result of a catalytic cyclization. Thus, upon exposition of [(3-iodoprop-2-yn-1-yl)oxy]arenes to IPrAuNTf2 (3 mol %), in 1,4-dioxane at 100 °C, the desired heterocyclic motif is readily assembled.
Macromolecules, 40, 1864-1864 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service