Skip to Content
MilliporeSigma

463752

Strontium hydroxide octahydrate

greener alternative

99.995% trace metals basis

Synonym(s):

Strontium dihydroxide octahydrate, Strontium hydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
Sr(OH)2 · 8H2O
CAS Number:
Molecular Weight:
265.76
NACRES:
NA.23
PubChem Substance ID:
UNSPSC Code:
12352302
EC Number:
242-367-1
MDL number:
Assay:
99.995% trace metals basis
Form:
crystals and lumps
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

99.995% trace metals basis

form

crystals and lumps

reaction suitability

core: strontium

greener alternative product characteristics

Design for Energy Efficiency
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product, Greener Alternative Product

impurities

≤55.0 ppm Trace Metal Analysis

density

1.9 g/mL at 25 °C (lit.)

greener alternative category

SMILES string

O.O.O.O.O.O.O.O.O[Sr]O

InChI

1S/10H2O.Sr/h10*1H2;/q;;;;;;;;;;+2/p-2

InChI key

UJPWWRPNIRRCPJ-UHFFFAOYSA-L

General description

We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives. This is an enabling product and can be used as a precursor to make perovskite oxides, which promote thermoelectric heat-to-electricity conversion, improving energy efficiency. Click here for more information.


Still not finding the right product?

Explore all of our products under Strontium hydroxide octahydrate


pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

supp_hazards

Storage Class

8A - Combustible corrosive hazardous materials

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library